Methyl oxalyl chloride 97%

Supplier: Thermo Scientific
Danger

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

166000250 166000500
ACRO166000250EA 121 EUR
ACRO166000250 ACRO166000500
Methyl oxalyl chloride 97%
Methyl oxalyl chloride
Formula: C₃H₃ClO₃
MW: 122,51 g/mol
Boiling Pt: 118…120 °C (760 mmHg)
Density: 1,33 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000705
CAS Number: 5781-53-3
EINECS: 227-307-4
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4179 to 1.4199 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤200 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR