Methyl oxalyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L13258.06
L13258.14
L13258.22
L13258.06EA
39.5
EUR
L13258.06
L13258.14
L13258.22
Methyl oxalyl chloride ≥97%
Methyl oxalyl chloride
Purity: 97%
CAS No.: 5781-53-3
Molecular Formula: C3H3ClO3
Formula Weight: 122.51
Flash Point: 46°(114°F)
Refractive Index: 1.4190
Odour: Acrid
Boiling Point: 115-118°
Beilstein Reference: 1071541
UN No.: 2920
MDL No.: MFCD00000705
Formula:
C₃H₃ClO₃ Boiling Pt: 115…118 °C Density: 1,332 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000705 CAS Number: 5781-53-3 EINECS: 227-307-4 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...