2-Chloro-5-ethylpyrimidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

H26856.06 H26856.14
H26856.06EA 87.6 EUR
H26856.06 H26856.14
2-Chloro-5-ethylpyrimidine ≥98%
2-Chloro-5-ethylpyrimidine

Purity: 98% CAS No.: 111196-81-7 Molecular Formula: C6H7ClN2 Formula Weight: 142.59 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.521 Boiling Point: 223° MDL No.: MFCD00799503

Formula: C₆H₇ClN₂
Boiling Pt: 223 °C
Density: 1,174
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00799503
CAS Number: 111196-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR