Methyl mercaptoacetate 95%

Supplier: Thermo Scientific
Danger

Synonyms: Methyl thioglycolate

148885000 148881000
ACRO148885000EA 93.8 EUR
ACRO148885000 ACRO148881000
Methyl mercaptoacetate 95%
Methyl mercaptoacetate
Formula: C₃H₆O₂S
MW: 106,15 g/mol
Boiling Pt: 156…159 °C (10 mmHg)
Melting Pt: –24 °C
Density: 1,187 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00004873
CAS Number: 2365-48-2
EINECS: 219-121-7
UN: 1228
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4670 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR