3-Methyl-1H-pyrazole 99%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Methylpyrazole

127560050 127560250
ACRO127560050EA 39.3 EUR
ACRO127560050 ACRO127560250
3-Methyl-1H-pyrazole 99%
3-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82,11 g/mol
Density: 1,02 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005240
CAS Number: 1453-58-3
EINECS: 215-925-7

Order Now

Specification Test Results

Appearance Clear colorless to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4945 to 1.4965 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR