4-Piperidinemethanol 98%

Supplier: Thermo Scientific
Danger

Synonyms: Piperidin-4-ylmethanol

380630050 380630250
ACRO380630050EA 96.4 EUR
ACRO380630050 ACRO380630250
4-Piperidinemethanol 98%
4-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115,18 g/mol
Boiling Pt: 118…120 °C (10 mmHg)
Melting Pt: 57…61 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00174228
CAS Number: 6457-49-4
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder and/or chunks or solid
Infrared spectrum Conforms
Melting point 55 to 62 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR