Ethyl butyrylacetate 98%

Supplier: Thermo Scientific

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

118190250 118191000
ACRO118190250EA 36 EUR
ACRO118190250 ACRO118191000
Ethyl butyrylacetate 98%
Ethyl butyrylacetate
Formula: C₈H₁₄O₃
MW: 158,2 g/mol
Boiling Pt: 104 °C (22 mmHg)
Density: 1 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009401
CAS Number: 3249-68-1
EINECS: 221-835-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4290 (20°C, 589 nm)
GC ≥97.5 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR