Chloromethyl chloroformate ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 97%
CAS No.: 22128-62-7
Molecular Formula: C2H2Cl2O2
Formula Weight: 128.94
Storage Temperature: 2°C to 8°C
Flash Point: 95°(203°F)
Refractive Index: 1.4295
Odour: Acrid
Melting Point:
Formula:
C₂H₂Cl₂O₂ Boiling Pt: 107…108 °C Melting Pt: < –20 °C Density: 1,465 Flash Pt: 95 °C (203 °F) |
MDL Number:
MFCD00077688 CAS Number: 22128-62-7 EINECS: 244-793-3 UN: 2745 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...