D-(-)-Ornithine hydrochloride ≥98%
Supplier: Thermo Scientific
Synonyms:
H-D-Orn-OH·HCl, H-D-Orn-OH.HCl
L00793.03
L00793.06
L00793.03EA
32.4
EUR
L00793.03
L00793.06
D-(-)-Ornithine hydrochloride ≥98%
D(-)-Ornithine hydrochloride
Purity: 98+%
CAS No.: 16682-12-5
Molecular Formula: C5H12N2O2.HCl
Formula Weight: 168.62
Melting Point: 237° dec.
Beilstein Reference: 4153339
Formula:
C₅H₁₂N₂O₂·HCl MW: 168,62 g/mol Melting Pt: 237 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012917 CAS Number: 16682-12-5 EINECS: 240-729-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...