Ethyl cyanoformate ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
L15777.14
L15777.06
L15777.22
L15777.14EA
139
EUR
L15777.14
L15777.06
L15777.22
Ethyl cyanoformate ≥99%
Ethyl cyanoformate
Purity: 99%
CAS No.: 623-49-4
Molecular Formula: NCCO2CH2CH3
Formula Weight: 99.09
Flash Point: 24°(75°F)
Refractive Index: 1.3820
Boiling Point: 115-116°
Beilstein Reference: 635955
UN No.: 3275
MDL No.: MFCD00001836
Formula:
C₄H₅NO₂ Boiling Pt: 115…116 °C Density: 1,003 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001836 CAS Number: 623-49-4 EINECS: 210-797-9 UN: 3275 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...