Ethyl isobutyrylacetate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 4-methyl-3-oxopentanoate
383440100
383440500
ACRO383440100EA
101
EUR
ACRO383440100
ACRO383440500
Ethyl isobutyrylacetate 95%
Ethyl isobutyrylacetate
Formula:
C₈H₁₄O₃ MW: 158,2 g/mol Boiling Pt: 173 °C (760 mmHg) Melting Pt: –9 °C Density: 0,981 g/cm³ Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009198 CAS Number: 7152-15-0 EINECS: 230-491-9 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4240 to 1.4280 (20°C, 589 nm) |
GC | ≥94 % (Mixture of tautomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...