6-Dodecyne ≥98%
Supplier: Thermo Fisher Scientific
L11262.06
L11262.14
L11262.06EA
66.5
EUR
L11262.06
L11262.14
6-Dodecyne ≥98%
6-Dodecyne
Purity: 98%
CAS No.: 6975-99-1
Molecular Formula: C12H22
Formula Weight: 166.31
Refractive Index: 1.4390
Boiling Point: 99-100°/4mm
MDL No.: MFCD00041658
Formula:
C₁₂H₂₂ MW: 166.31 g/mol Boiling Pt: 99…100 °C (4 mmHg) Density: 0.790 Storage Temperature: Ambient |
MDL Number:
MFCD00041658 CAS Number: 6975-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...