4-Amino-1-benzylpiperidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Amino-N-benzylpiperidine

A10149.14
A10149.14EA 0 EUR
A10149.14
4-Amino-1-benzylpiperidine ≥98%
4-Amino-1-benzylpiperidine

Purity: 98% CAS No.: 50541-93-0 Molecular Formula: C12H18N2 Formula Weight: 190.29 Flash Point: 110°(230°F) Refractive Index: 1.5430 Boiling Point: 279-281° Beilstein Reference: 146038 MDL No.: MFCD00006504

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 279…281 °C
Density: 0.930
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006504
CAS Number: 50541-93-0
EINECS: 256-620-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR