(Phenylthio)acetonitrile ≥98%

Supplier: Thermo Fisher Scientific
Warning

A12869.14 A12869.22
A12869.14EA 118 EUR
A12869.14 A12869.22
(Phenylthio)acetonitrile ≥98%
(Phenylthio)acetonitrile

Purity: 98% CAS No.: 5219-61-4 Molecular Formula: C6H5SCH2CN Formula Weight: 149.22 Flash Point: 110°(230°F) Refractive Index: 1.5820 Odour: Unpleasant Boiling Point: 146-147°/14mm Beilstein Reference: 2042078 MDL No.: MFCD00019831

Formula: C₈H₇NS
Boiling Pt: 146…147 °C (14 mmHg)
Density: 1,142
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019831
CAS Number: 5219-61-4
EINECS: 226-013-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR