Trimellitic anhydride 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,2,4-Benzenetricarboxylic anhydride, 1,2,4-Benzenetricarboxylic 1,2-anhydride

105370010 105372500
ACRO105370010EA 86 EUR
ACRO105370010 ACRO105372500
Trimellitic anhydride 97%
Trimellitic anhydride
Formula: C₉H₄O₅
MW: 192.13 g/mol
Melting Pt: 164…169 °C
Flash Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD00005925
CAS Number: 552-30-7
EINECS: 209-008-0

Order Now

Specification Test Results

Appearance White to off-white Flakes
Infrared spectrum Conforms
Melting point 164 to 169 °C
Assay ≥96.0 %
Absorption Ratio P-15/P+15 0.98 to 1.20
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR