Pyromellitic dianhydride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1,2,4,5-Benzenetetracarboxylic anhydride
45788.A3
45788.22
45788.36
45788.A3EA
979
EUR
45788.A3
45788.22
45788.36
Pyromellitic dianhydride ≥98%
Pyromellitic dianhydride
Purity: 98%
CAS No.: 89-32-7
Molecular Formula: C10H2O6
Formula Weight: 218.12
Flash Point: 380°(716°F)
Physical Form: Crystalline powder
Density: 1.68
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 284-288°
Boiling Point: 305-310°/30mm
Beilstein Reference: 213583
MDL No.: MFCD00005005
Formula:
C₁₀H₂O₆ MW: 218,12 g/mol Boiling Pt: 305…310 °C (30 mmHg) Melting Pt: 284…288 °C Density: 1,68 Flash Pt: 380 °C (716 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005005 CAS Number: 89-32-7 EINECS: 201-898-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...