Pyromellitic dianhydride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1,2,4,5-Benzenetetracarboxylic anhydride
A12712.0E
A12712.22
A12712.36
A12712.0EEA
632
EUR
A12712.0E
A12712.22
A12712.36
Pyromellitic dianhydride ≥97%
Pyromellitic dianhydride
Purity: 97%
CAS No.: 89-32-7
Molecular Formula: C10H2O6
Formula Weight: 218.12
Flash Point: 380°(716°F)
Melting Point: 284-288°
Boiling Point: 305-310°/30mm
Beilstein Reference: 213583
MDL No.: MFCD00005005
Formula:
C₁₀H₂O₆ Boiling Pt: 305…310 °C (30 mmHg) Melting Pt: 284…288 °C Density: 1,68 Flash Pt: 380 °C (716 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005005 CAS Number: 89-32-7 EINECS: 201-898-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...