2,4-Thiazolidinedione 99%

Supplier: Thermo Scientific
Warning

Synonyms: 1,3-Thiazolidine-2,4-dione

159490250 159491000
ACRO159490250EA 79 EUR
ACRO159490250 ACRO159491000
2,4-Thiazolidinedione 99%
2,4-Thiazolidinedione
Formula: C₃H₃NO₂S
MW: 117,13 g/mol
Boiling Pt: 178…179 °C (19 mmHg)
Melting Pt: 123…126 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00005478
CAS Number: 2295-31-0
EINECS: 218-941-2

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
HPLC ≥98.5 %
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR