tert-Butyl hydroperoxide ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
TBHP
M-Clarity™ quality level = MQ200
8.14006.0005
8.14006.0250
8.14006.0005EA
38.1
EUR
8.14006.0005
8.14006.0250
tert-Butyl hydroperoxide ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
tert-Butyl hydroperoxide
Formula:
C₄H₁₀O₂ MW: 90,12 g/mol Density: 0,94 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002130 CAS Number: 75-91-2 EINECS: 200-915-7 UN: 3109 ADR: 5.2 (8), |
Specification Test Results
Solubility | 130 - 150 g/l (20 °C) |
Melting Point | -3 °C |
Boiling Point | 37 °C (20 hPa) |
Vapor Pressure | 232 hPa (60 °C) |
Flash Point | 38 °C |
Refractive Index | 1.4015 (20 °C, 589 nm) |
Density | 0.94 g/cm3 (20 °C) |
Hazard Class | 5.2 |
Hazard Class Description | Organic peroxide |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...