Norbornadiene ≥97% stabilised

Supplier: Thermo Scientific
Danger

Synonyms: 2,5-Norbornadiene

Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

L13935.AC L13935.AE
L13935.ACEA 47.2 EUR
L13935.AC L13935.AE
Norbornadiene ≥97% stabilised
Norbornadiene

Stabilized with 250 ppm BHT


Purity: 97% CAS No.: 121-46-0 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: -11°(12°F) Refractive Index: 1.4700 Melting Point: -20° Boiling Point: 88-90° Beilstein Reference: 506224 UN No.: 2251 MDL No.: MFCD00082301

Formula: C₇H₈
MW: 92,14 g/mol
Boiling Pt: 88…90 °C
Melting Pt: –20 °C
Density: 0,906
Flash Pt: –11 °C (–12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082301
CAS Number: 121-46-0
EINECS: 204-472-0
UN: 2251
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR