2-Benzoxazolinone 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
benzo[d]oxazol-2(3H)-one
169080250
169081000
169085000
ACRO169080250EA
58.3
EUR
ACRO169080250
ACRO169081000
ACRO169085000
2-Benzoxazolinone 98%
2-Benzoxazolinone
Formula:
C₇H₅NO₂ MW: 135,12 g/mol Melting Pt: 137…141 °C Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005716 CAS Number: 59-49-4 EINECS: 200-430-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Light beige to grey-brown Powder |
Infrared spectrum | Conforms |
Melting point | 137 to 141 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...