2,5-Dimercapto-1,3,4-thiadiazole, (max. 3% H₂O) 97% (dry weight)

Supplier: Thermo Scientific
Warning

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

A12615.18 A12615.30
A12615.18EA 50.6 EUR
A12615.18 A12615.30
2,5-Dimercapto-1,3,4-thiadiazole, (max. 3% H₂O) 97% (dry weight)
2,5-Dimercapto-1,3,4-thiadiazole

Purity: 98% CAS No.: 1072-71-5 Molecular Formula: C2H2N2S3 Formula Weight: 150.24 Melting Point: ca 162° dec. Beilstein Reference: 113444

Formula: C₂H₂N₂S₃
MW: 150,25 g/mol
Melting Pt: ∼ 162 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003103
CAS Number: 1072-71-5
EINECS: 214-014-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR