Biocide, concentrate, sterile, filtered, amine unit dose, Klercide®
Supplier: Ecolab
SHIE3078720EA 0 EUR
SHIE3078720
Biocide, concentrate, sterile, filtered, amine unit dose, Klercide®
Alcohol-free, amphoteric surfactant biocide designed for cleanroom use. Filtered to 0,2 µm and filled in an ISO Class 7 Grade C cleanroom before irradiation. Five bottles are double bagged.
  • Convenient unit dose saves time and avoids dilution errors
  • CEN and isolate tested
  • Safe and effective; non toxic and non corrosive
  • Fully compatible and ideal for a rotational regime
Caution: Use biocides safely. Always read the label and product information before use.