Indole-7-carboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H54931.03
H54931.06
H54931.14
H54931.03EA
45.7
EUR
H54931.03
H54931.06
H54931.14
Indole-7-carboxylic acid ≥97%
Indole-7-carboxylic acid
Purity: 97%
CAS No.: 1670-83-3
Molecular Formula: C9H7NO2
Formula Weight: 161.05
MDL No.: MFCD00210442
Formula:
C₉H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00210442 CAS Number: 1670-83-3 EINECS: 216-801-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...