2-Nitrofluorene 98
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Nitro-9H-fluorene
H55744.06
H55744.14
H55744.06EA
44.5
EUR
H55744.06
H55744.14
2-Nitrofluorene 98
2-Nitrofluorene
Formula:
C₁₃H₉NO₂ MW: 211.22 g/mol Melting Pt: 156…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001117 CAS Number: 607-57-8 EINECS: 210-138-5 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...