2,7-Diaminofluorene 97%

Supplier: Thermo Scientific
Danger

Synonyms: Fluorene-2,7-diamine, 2,7-Diamino-fluorene, Fluorene-2,7-diyldiamine

H55402.03 H55402.06
H55402.03EA 92.4 EUR
H55402.03 H55402.06
2,7-Diaminofluorene 97%
2,7-Diaminofluorene

Purity: 97+% CAS No.: 525-64-4 Molecular Formula: C13H12N2 Formula Weight: 196.25 Melting Point: 160-162° Beilstein Reference: 2099859 Merck Reference: 14,4156 MDL No.: MFCD00001128

Formula: C₁₃H₁₂N₂
MW: 196,25 g/mol
Boiling Pt: 467 °C (1013 hPa)
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00001128
CAS Number: 525-64-4
EINECS: 208-377-5
Merck Index: 14,4156

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR