2,4,6-Triphenoxy-1,3,5-triazine 96%

Supplier: Thermo Scientific
Danger

Synonyms: 2,4,6-Triphenoxy-s-triazine

H33572.09 H33572.18
H33572.09EA 0 EUR
H33572.09 H33572.18
2,4,6-Triphenoxy-1,3,5-triazine 96%
2,4,6-Triphenoxy-1,3,5-triazine

Purity: 96% CAS No.: 1919-48-8 Molecular Formula: C21H15N3O3 Formula Weight: 357.36 Melting Point: 235-236° MDL No.: MFCD00006048

Formula: C₂₁H₁₅N₃O₃
MW: 357,37 g/mol
Melting Pt: 235…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00006048
CAS Number: 1919-48-8
EINECS: 217-644-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR