4-Methoxysalicylic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H55345.06 H55345.14
H55345.06EA 57 EUR
H55345.06 H55345.14
4-Methoxysalicylic acid ≥98%
4-Methoxysalicylic acid

Purity: 98% CAS No.: 2237-36-7 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 158-159° MDL No.: MFCD00002450

Formula: C₈H₈O₄
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 158…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002450
CAS Number: 2237-36-7
EINECS: 218-801-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR