1-(4-Chlorobenzyl)piperazine ≥98%

Supplier: Thermo Fisher Scientific
Danger

H55337.03 H55337.06 H55337.14
H55337.03EA 35.4 EUR
H55337.03 H55337.06 H55337.14
1-(4-Chlorobenzyl)piperazine ≥98%
1-(4-Chlorobenzyl)piperazine
Formula: C₁₁H₁₅ClN₂
MW: 210.71 g/mol
Boiling Pt: 104…106 °C (0.1 mmHg)
Density: 1.140
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040791
CAS Number: 23145-88-2
UN: 3267
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR