Methyl picolinate ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl pyridine-2-carboxylate
H54530.14
H54530.22
H54530.14EA
128
EUR
H54530.14
H54530.22
Methyl picolinate ≥99%
Methyl picolinate
Purity: 99%
CAS No.: 2459-07-6
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 103°(217°F)
Refractive Index: 1.521
Melting Point: 19°
Boiling Point: 95°/1mm
MDL No.: MFCD00038038
Formula:
C₇H₇NO₂ MW: 137,14 g/mol Boiling Pt: 95 °C (1 mmHg) Melting Pt: 19 °C Density: 1,137 Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038038 CAS Number: 2459-07-6 EINECS: 219-545-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...