3-Dodecylthiophene ≥97%

Supplier: Thermo Fisher Scientific

H55799.03 H55799.06
H55799.03EA 64 EUR
H55799.03 H55799.06
3-Dodecylthiophene ≥97%
3-Dodecylthiophene

Purity: 97% CAS No.: 104934-52-3 Molecular Formula: C16H28S Formula Weight: 252.46 Refractive Index: 1.4880 Boiling Point: 290° MDL No.: MFCD00130121

Formula: C₁₆H₂₈S
Boiling Pt: 290 °C
Density: 0,902
Storage Temperature: Ambient
MDL Number: MFCD00130121
CAS Number: 104934-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR