3-Hexylthiophene 99

Supplier: Thermo Fisher Scientific
Warning

H55891.03 H55891.06 H55891.14
H55891.03EA 39.3 EUR
H55891.03 H55891.06 H55891.14
3-Hexylthiophene 99
3-Hexylthiophene
Formula: C₁₀H₁₆S
MW: 168.3 g/mol
Boiling Pt: 65 °C (0.45 mmHg)
Density: 0.936
Flash Pt: 37 °C (99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00143181
CAS Number: 1693-86-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR