Malonyl dichloride 97%
Supplier: Thermo Scientific
|
Danger
|
125280250
125281000
ACRO125280250EA
103
EUR
ACRO125280250
ACRO125281000
Malonyl dichloride 97%
Malonyl dichloride
Formula:
C₃H₂Cl₂O₂ MW: 140,95 g/mol Boiling Pt: 47 °C (15 mmHg) Density: 1,45 g/cm³ Flash Pt: 47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000735 CAS Number: 1663-67-8 EINECS: 216-772-9 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow to orange to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4620 to 1.4660 (20°C, 589 nm) |
Titration Argentometric | ≥96.0 % |
Residue after evaporation | ≤0.0003 % |
Conductivity | ≤1 µS/cm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...