5-Bromo-1-tetralone
Supplier: Cayman Chemical
Synonyms:
5-Bromo-3,4-dihydronaphthalen-1(2H)-one
10007134-250
10007134-500
CAYM10007134-250EA
74.5
EUR
CAYM10007134-250
CAYM10007134-500
5-Bromo-1-tetralone
5-Bromo-1-tetralone
Formula:
C₁₀H₉BrO MW: 225,08 g/mol |
MDL Number:
MFCD02179288 CAS Number: 68449-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...