7-Nitro-1-tetralone ≥97%
Supplier: Thermo Scientific
Synonyms:
3,4-Dihydro-7-nitronaphthalen-1(2H)-one, 7-Nitro-3,4-dihydronaphthalen-1(2H)-one
L01853.04
L01853.09
L01853.04EA
85.8
EUR
L01853.04
L01853.09
7-Nitro-1-tetralone ≥97%
7-Nitro-1-tetralone
Purity: 97%
CAS No.: 40353-34-2
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Melting Point: 103-108°
Beilstein Reference: 1570515
Formula:
C₁₀H₉NO₃ MW: 191,19 g/mol Melting Pt: 103…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019661 CAS Number: 40353-34-2 EINECS: 254-887-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...