7-Methoxy-1-tetralone ≥99%
Supplier: Thermo Fisher Scientific
Synonyms:
7-Methoxy-3,4-dihydronaphthalen-1(2H)-one
L15130.03
L15130.06
L15130.03EA
0
EUR
L15130.03
L15130.06
7-Methoxy-1-tetralone ≥99%
7-Methoxy-1-tetralone
Purity: 99%
CAS No.: 6836-19-7
Molecular Formula: C11H12O2
Formula Weight: 176.22
Melting Point: 59-63°
Beilstein Reference: 2208389
Formula:
C₁₁H₁₂O₂ Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001696 CAS Number: 6836-19-7 EINECS: 229-916-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...