3-Chloro-6-hydrazinopyridazine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Chloro-6-hydrazinylpyridazine

L06121.03 L06121.06
L06121.03EA 23.4 EUR
L06121.03 L06121.06
3-Chloro-6-hydrazinopyridazine ≥98%
3-Chloro-6-hydrazinopyridazine

Purity: 98% CAS No.: 17284-97-8 Molecular Formula: C4H5ClN4 Formula Weight: 144.56 Melting Point: 134-140° Beilstein Reference: 125591

Formula: C₄H₅ClN₄
MW: 144,56 g/mol
Melting Pt: 134…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00051740
CAS Number: 17284-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR