2-Bromo-4-methylpyrimidine ≥97%

Supplier: Thermo Scientific
Warning

H54503.03 H54503.06 H54503.14
H54503.03EA 89.2 EUR
H54503.03 H54503.06 H54503.14
2-Bromo-4-methylpyrimidine ≥97%
2-Bromo-4-methylpyrimidine

Purity: 97% CAS No.: 130645-48-6 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 45-50° MDL No.: MFCD04037883

Formula: C₅H₅ClN₂
Melting Pt: 45…50 °C
Storage Temperature: Ambient
MDL Number: MFCD04037883
CAS Number: 130645-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR