1,4-Dithiane-2,5-diol 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol
A14620.0B
A14620.18
A14620.30
A14620.0BEA
324
EUR
A14620.0B
A14620.18
A14620.30
1,4-Dithiane-2,5-diol 96%
1,4-Dithiane-2,5-diol
Purity: 96%
CAS No.: 40018-26-6
Molecular Formula: C4H8O2S2
Formula Weight: 152.24
Storage Temperature: 2°C to 8°C
Melting Point: ca 130° dec.
Beilstein Reference: 104402
UN No.: 2811
MDL No.: MFCD00006659
Formula:
C₄H₈O₂S₂ MW: 152,24 g/mol Melting Pt: ∼ 130 °C (dec.) |
MDL Number:
MFCD00006659 CAS Number: 40018-26-6 EINECS: 254-751-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...