Nitrilotriacetic acid trisodium salt monohydrate 99+%
Supplier: Thermo Scientific
|
Warning
|
147840250
147845000
147840025
ACRO147840250EA
35.4
EUR
ACRO147840250
ACRO147845000
ACRO147840025
Nitrilotriacetic acid trisodium salt monohydrate 99+%
Nitrilotriacetic acid trisodium salt monohydrate
Formula:
C₆H₆NNa₃O₆·1H₂O Melting Pt: >320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150789 CAS Number: 18662-53-8 EINECS: 225-768-6 |
Specification Test Results
Appearance | White Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...