trans-Aconitic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid
B20087.14
B20087.22
B20087.PG
B20087.14EA
70.4
EUR
B20087.14
B20087.22
B20087.PG
trans-Aconitic acid ≥98%
trans-Aconitic acid
Purity: 98%
CAS No.: 4023-65-8
Molecular Formula: C6H6O6
Formula Weight: 174.11
Odour: Odourless
Melting Point: ca 190° dec.
MDL No.: MFCD00002721
Formula:
C₆H₆O₆ MW: 174,11 g/mol Melting Pt: ∼ 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002721 CAS Number: 4023-65-8 EINECS: 223-688-6 Merck Index: 14,117 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...