2-Chloro-6-hydrazinopyridine 95%
|
Warning
|
Synonyms:
(6-Chloro-2-pyridyl)hydrazine
H27249.MD
H27249.03
H27249.MDEA
98.4
EUR
H27249.MD
H27249.03
2-Chloro-6-hydrazinopyridine 95%
2-Chloro-6-hydrazinopyridine
Purity: 95%
CAS No.: 5193-03-3
Molecular Formula: C5H6ClN3
Formula Weight: 143.58
Boiling Point: 117-120°
MDL No.: MFCD00214724
Formula:
C₅H₆ClN₃ Boiling Pt: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00214724 CAS Number: 5193-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...