(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98%

Supplier: Thermo Scientific

Synonyms: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

L14518.03 L14518.MD
L14518.03EA 120 EUR
L14518.03 L14518.MD
(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98%
(R)-(+)-4-Isopropyl-2-oxazolidinone

Purity: 98+% CAS No.: 95530-58-8 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: 70-74° Beilstein Reference: 5376658

Formula: C₆H₁₁NO₂
MW: 129,16 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00075172
CAS Number: 95530-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR