(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (R)-1,2,3,4-Tetrahydronaphthalen-1-amine

L17021.06 L17021.03
L17021.06EA 0 EUR
L17021.06 L17021.03
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 98%, ChiPros®
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine

Purity: 99+% CAS No.: 23357-46-2 Molecular Formula: C10H13N Formula Weight: 147.22 Flash Point: 110°(230°F) Refractive Index: 1.5645 Boiling Point: 118-120°/8mm Beilstein Reference: 4231302 MDL No.: MFCD00671629

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 118…120 °C (8 mmHg)
Density: 1.035
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671629
CAS Number: 23357-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR