4-tert-Butylcalix[4]arene 99%
Supplier: Thermo Fisher Scientific
Synonyms:
p-tert-Butylcalix[4]arene
290220250
290220050
ACRO290220250EA
252
EUR
ACRO290220250
ACRO290220050
4-tert-Butylcalix[4]arene 99%
4-tert-Butylcalix[4]arene
Formula:
C₄₄H₅₆O₄ MW: 648.93 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066280 CAS Number: 60705-62-6 |
Specification Test Results
Appearance | White to light beige Crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...