4-Cumylphenol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23410.0B
B23410.22
B23410.30
B23410.0BEA
242
EUR
B23410.0B
B23410.22
B23410.30
4-Cumylphenol ≥97%
4-Cumylphenol
Purity: 97%
CAS No.: 599-64-4
Molecular Formula: C15H16O
Formula Weight: 212.29
Flash Point: 160°(320°F)
Melting Point: 70-73°
Boiling Point: 189-191°/10mm
Beilstein Reference: 1870517
MDL No.: MFCD00002365
Formula:
C₁₅H₁₆O Boiling Pt: 189…191 °C (10 mmHg) Melting Pt: 70…73 °C Density: 1,115 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002365 CAS Number: 599-64-4 EINECS: 209-968-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...