4-Nonyloxybenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
L09110.09
L09110.04
L09110.09EA
0
EUR
L09110.09
L09110.04
4-Nonyloxybenzaldehyde ≥98%
4-Nonyloxybenzaldehyde
Purity: 98%
CAS No.: 50262-46-9
Molecular Formula: C16H24O2
Formula Weight: 248.37
Refractive Index: 1.5190
Boiling Point: 180-182°/4mm
MDL No.: MFCD00043712
Formula:
C₁₆H₂₄O₂ MW: 248.37 g/mol Boiling Pt: 180…182 °C (4 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00043712 CAS Number: 50262-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...