2-Bromo-5-methoxyphenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Bromo-5-methoxybenzeneboronic acid, 2-Bromo-5-methoxybenzene boronic acid

H52972.MD H52972.03
H52972.MDEA 0 EUR
H52972.MD H52972.03
2-Bromo-5-methoxyphenylboronic acid ≥97%
2-Bromo-5-methoxyphenylboronic acid

Purity: 97% CAS No.: 89694-44-0 Molecular Formula: C7H8BBrO3 Formula Weight: 230.85 Melting Point: 214-216° MDL No.: MFCD06659859

Formula: C₇H₈BBrO₃
MW: 230,85 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD06659859
CAS Number: 89694-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR