2-Chloro-4-methoxyphenylboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-4-methoxybenzeneboronic acid

H26917.06 H26917.03
H26917.06EA 311 EUR
H26917.06 H26917.03
2-Chloro-4-methoxyphenylboronic acid 95%
2-Chloro-4-methoxyphenylboronic acid

Purity: 95% CAS No.: 219735-99-6 Molecular Formula: C7H8BClO3 Formula Weight: 186.40 Melting Point: ca 202° MDL No.: MFCD03411935

Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Melting Pt: ∼ 202 °C
Storage Temperature: Ambient
MDL Number: MFCD03411935
CAS Number: 219735-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR