3-Bromophenylurea ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
1-(3-Bromophenyl)urea
L11370.03
L11370.06
L11370.03EA
31.4
EUR
L11370.03
L11370.06
3-Bromophenylurea ≥97%
3-Bromophenylurea
Purity: 97%
CAS No.: 2989-98-2
Molecular Formula: C7H7BrN2O
Formula Weight: 215.06
Melting Point: 166-168°
Beilstein Reference: 2718463
Formula:
C₇H₇BrN₂O MW: 215.06 g/mol Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041317 CAS Number: 2989-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...